Learn More
Medchemexpress LLC HY-N1984 5mg Medchemexpress, Artemisic acid CAS:80286-58-4 Purity:>98%
SDP
Catalog No. 501970354
Encompass_Preferred
Medchemexpress, HY-N1984 5mg Artemisic acid CAS:80286-58-4 Artemisinic acid (Qing Hao acid), an amorphane sesquiterpene isolated from Artemisia annua L., possesses a variety of pharmacological activity, such as antimalarial activity, anti-tumor activity, antipyretic effect, antibacterial activity, allelopathy effect and anti-adipogenesis effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0354 Supplier Medchemexpress LLC Supplier No. HYN19845MG
Medchemexpress, HY-N1984 5mg Artemisic acid CAS:80286-58-4 Artemisinic acid (Qing Hao acid), an amorphane sesquiterpene isolated from Artemisia annua L., possesses a variety of pharmacological activity, such as antimalarial activity, anti-tumor activity, antipyretic effect, antibacterial activity, allelopathy effect and anti-adipogenesis effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.