Learn More
Cayman Chemical ArtemIsInIc AcId 10mg
SDP
Catalog No. 502933951
Encompass_Preferred
A sesquiterpene; decreases triglyceride levels and GPDH activity in hAMSCs; inhibits adipocyte differentiation at 200 µM; decreases C/EBPα and HMG-CoA reductase mRNA expression and inhibits cholesterol synthesis and melanogenesis in human epidermal melanocytes
Catalog No. 50-293-3951 Supplier Cayman Chemical Supplier No. 2505910 MG
A sesquiterpene; decreases triglyceride levels and GPDH activity in hAMSCs; inhibits adipocyte differentiation at 200 µM; decreases C/EBPα and HMG-CoA reductase mRNA expression and inhibits cholesterol synthesis and melanogenesis in human epidermal melanocytes