Learn More
Cayman Chemical ArtemIsInIc AcId 50mg
SDP
Catalog No. 502935188
Encompass_Preferred
A sesquiterpene; decreases triglyceride levels and GPDH activity in hAMSCs; inhibits adipocyte differentiation at 200 µM; decreases C/EBPα and HMG-CoA reductase mRNA expression and inhibits cholesterol synthesis and melanogenesis in human epidermal melanocytes
Catalog No. 50-293-5188 Supplier Cayman Chemical Supplier No. 2505950 MG
A sesquiterpene; decreases triglyceride levels and GPDH activity in hAMSCs; inhibits adipocyte differentiation at 200 µM; decreases C/EBPα and HMG-CoA reductase mRNA expression and inhibits cholesterol synthesis and melanogenesis in human epidermal melanocytes