Learn More
Enzo Life Sciences Artemisinin (100mg). CAS: 63968-64-9
SDP
Supplier:  Enzo Life Sciences ALX350219M100
Encompass_Preferred
Powerful antimalarial agent. Inhibits angiogenesis by down-regulating HIF-1α and VEGF expression in mouse embryonic stem cells. Crosses the blood-brain barrier. Inhibitor of human iNOS (NOS II). Alternative name: Qinghaosu. Purity: ≥98% (TLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in methanol, 100% ethanol, dimethyl formamide, DMSO or acetone; almost insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8783