Learn More
AdipoGen Artemisinin (100 mg)
SDP
Supplier:  AdipoGen AGCN20467M100
Encompass_Preferred
Chemical. CAS: 63968-64-9. Formula: C15H22O5. MW: 282.3. Isolated from Artemisia annua. Anti-malaria. Apoptosis inducer. Anti-cancer. Inhibits production of tumor necrosis factor-alpha (TNF-alpha), interleukin-6 (IL-6), monocyte chemotactic protein-1 (MCP-1) and nitric oxide (NO).
Catalog No. 50-205-3643