Learn More
Selleck Chemical LLC Artemisinin S1282-50mg
SDP
Catalog No. 503108110
Encompass_Preferred
Artemisinin is a sesquiterpene endoperoxide which is a potent antimalarial agent
Catalog No. 50-310-8110 Supplier Selleck Chemical LLC Supplier No. S128250MG
Artemisinin is a sesquiterpene endoperoxide which is a potent antimalarial agent