Learn More
Cayman Chemical ArtemIsInIn 500mg
SDP
Catalog No. 502951928
Encompass_Preferred
An antimalarial agent with anticancer activity; an iron(II) oxide-reactive endoperoxide that generates ROS; reduces the growth of various P. falciparum strains in vitro (IC50s = 3.98-20.36 nM); reduces parasitemia in mice infected with P. falciparum (CD50 = 140 mg/kg); reduces P. berghei infection in mice (ED50 = 5.6 mg/kg per day); inhibits growth of SK-N-AS, BE(2)-C, SK-N-DZ, and SHEP1 neuroblastoma cells from 100-400 μM; suppresses BE(2)-C cell colony formation in a soft agar assay and reduces tumor growth in a BE(2)-C mouse xenograft model
Catalog No. 50-295-1928 Supplier Cayman Chemical Supplier No. 11816500 MG
An antimalarial agent with anticancer activity; an iron(II) oxide-reactive endoperoxide that generates ROS; reduces the growth of various P. falciparum strains in vitro (IC50s = 3.98-20.36 nM); reduces parasitemia in mice infected with P. falciparum (CD50 = 140 mg/kg); reduces P. berghei infection in mice (ED50 = 5.6 mg/kg per day); inhibits growth of SK-N-AS, BE(2)-C, SK-N-DZ, and SHEP1 neuroblastoma cells from 100-400 μM; suppresses BE(2)-C cell colony formation in a soft agar assay and reduces tumor growth in a BE(2)-C mouse xenograft model