Learn More
Medchemexpress LLC Artemisinin | 63968-64-9 | 98.0% | C15H22O5 | 1 G
SDP
Catalog No. 5000432834
Encompass_Preferred
Artemisinin (Qinghaosu), a sesquiterpene lactone, is an anti-malarial agent isolated from the aerial parts of Artemisia annua L. plants. It inhibits the AKT signaling pathway by decreasing pAKT in a dose-dependent manner. Artemisinin also reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis, and has neuroprotective effects.
  • Isolated from Artemisia annua L. plants.
  • Inhibits AKT signaling pathway.
  • Reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis.
  • Provides neuroprotective effects.
  • Acts as an anti-malarial agent.
Catalog No. 50-004-32834 Supplier Medchemexpress LLC Supplier No. HYB00941G
Artemisinin (Qinghaosu), a sesquiterpene lactone, is an anti-malarial agent isolated from the aerial parts of Artemisia annua L. plants. It inhibits the AKT signaling pathway by decreasing pAKT in a dose-dependent manner. Artemisinin also reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis, and has neuroprotective effects.
  • Isolated from Artemisia annua L. plants.
  • Inhibits AKT signaling pathway.
  • Reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis.
  • Provides neuroprotective effects.
  • Acts as an anti-malarial agent.