Learn More
Medchemexpress LLC Artemisinin | 63968-64-9 | 98.0% | C15H22O5 | 500 MG
SDP
Catalog No. 5000432633
Encompass_Preferred
Artemisinin, also known as Qinghaosu, is a sesquiterpene lactone and an anti-malarial agent derived from *Artemisia annua L.* plants. It inhibits the AKT signaling pathway, leading to a dose-dependent decrease in pAKT levels. Artemisinin reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis, and demonstrates neuroprotective effects.
  • Anti-malarial agent
  • Inhibits AKT signaling pathway
  • Reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis
  • Demonstrates neuroprotective capabilities
  • Targets Plasmodium
  • Exhibits antiproliferative activity against various human cancer cell lines
  • Shows antiparasitic activity against *Toxoplasma gondii*
  • Exhibits antiviral activity against Hepatitis B virus
Catalog No. 50-004-32633 Supplier Medchemexpress LLC Supplier No. HYB0094500MG
Artemisinin, also known as Qinghaosu, is a sesquiterpene lactone and an anti-malarial agent derived from *Artemisia annua L.* plants. It inhibits the AKT signaling pathway, leading to a dose-dependent decrease in pAKT levels. Artemisinin reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis, and demonstrates neuroprotective effects.
  • Anti-malarial agent
  • Inhibits AKT signaling pathway
  • Reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis
  • Demonstrates neuroprotective capabilities
  • Targets Plasmodium
  • Exhibits antiproliferative activity against various human cancer cell lines
  • Shows antiparasitic activity against *Toxoplasma gondii*
  • Exhibits antiviral activity against Hepatitis B virus