Learn More
Abcam Artemisinin, Antimalarial compound, 100MG
Catalog No. 502706694
Encompass_Preferred
MW 282.33 Da, Purity >99%. Antimalarial compound. Forms highly alkylating free radicals and reactive oxygen species (ROS°) on interaction with iron. Additionally shows anticancer and antitumor properties. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6694 Supplier Abcam Supplier No. AB141308100MG
MW 282.33 Da, Purity >99%. Antimalarial compound. Forms highly alkylating free radicals and reactive oxygen species (ROS°) on interaction with iron. Additionally shows anticancer and antitumor properties. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement