Learn More
Cayman Chemical Artemisinin-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 176652-07-6, 1 mg
SDP
Catalog No. 502932732
Encompass_Preferred
An internal standard for the quantification of artemisinin by GC- or LC-MS
Catalog No. 50-293-2732 Supplier Cayman Chemical Supplier No. 253591 MG
An internal standard for the quantification of artemisinin by GC- or LC-MS