Learn More
Medchemexpress LLC Artemisinin (Standard) | 63968-64-9 | 98.3% | C₁₅H₂₂O₅ | 250 MG
SDP
Catalog No. 5000358514
Encompass_Preferred
Artemisinin (Qinghaosu) is a sesquiterpene lactone and an anti-malarial agent isolated from the aerial parts of Artemisia annua L. plants. It inhibits the AKT signaling pathway by decreasing pAKT in a dose-dependent manner. Artemisinin reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis, and also has neuroprotective effects.
  • Sesquiterpene lactone.
  • Anti-malarial agent.
  • Inhibits AKT signaling pathway.
  • Reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis.
  • Has neuroprotective effects.
  • Used as an analytical standard for research experiments, including HPLC, GC, and MS.
Catalog No. 50-003-58514 Supplier Medchemexpress LLC Supplier No. 250MGHYB0094R
Artemisinin (Qinghaosu) is a sesquiterpene lactone and an anti-malarial agent isolated from the aerial parts of Artemisia annua L. plants. It inhibits the AKT signaling pathway by decreasing pAKT in a dose-dependent manner. Artemisinin reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis, and also has neuroprotective effects.
  • Sesquiterpene lactone.
  • Anti-malarial agent.
  • Inhibits AKT signaling pathway.
  • Reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis.
  • Has neuroprotective effects.
  • Used as an analytical standard for research experiments, including HPLC, GC, and MS.