Learn More
Medchemexpress LLC Artemisinin (Standard) | 63968-64-9 | 98.6% | C₁₅H₂₂O₅ | 100 MG
SDP
Catalog No. 5000432764
Encompass_Preferred
Artemisinin (Standard) is the analytical standard of Artemisinin, a sesquiterpene lactone and anti-malarial agent isolated from the aerial parts of Artemisia annua L. plants. It inhibits the AKT signaling pathway and reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis, also exhibiting neuroprotective effects. Intended for research and analytical applications, it is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard of artemisinin
  • Anti-malarial agent
  • Inhibits AKT signaling pathway
  • Reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis
  • Has neuroprotective effects
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-004-32764 Supplier Medchemexpress LLC Supplier No. HYB0094R100MG
Artemisinin (Standard) is the analytical standard of Artemisinin, a sesquiterpene lactone and anti-malarial agent isolated from the aerial parts of Artemisia annua L. plants. It inhibits the AKT signaling pathway and reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis, also exhibiting neuroprotective effects. Intended for research and analytical applications, it is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard of artemisinin
  • Anti-malarial agent
  • Inhibits AKT signaling pathway
  • Reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis
  • Has neuroprotective effects
  • Used in HPLC, GC, and MS experiments