Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Artemisinin (Standard) | 63968-64-9 | 98.6% | C₁₅H₂₂O₅ | 25 MG
SDP

Catalog No. 5000357845
Encompass_Preferred

Artemisinin (Standard) | 63968-64-9 | 98.6% | C₁₅H₂₂O₅ | 25 MG

Catalog No. 50-003-57845 Supplier Medchemexpress LLC Supplier No. 25MGHYB0094R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Artemisinin (Standard) | 63968-64-9 | 98.6% | C₁₅H₂₂O₅ | 25 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.