Learn More
Apexbio Technology LLC Artemisinine 63968-64-9 200mg
SDP
Catalog No. 5000090369
Encompass_Preferred
Artemisinine (CAS 63968-64-9) is a sesquiterpene lactone designed to modulate parasite cell integrity and cancer cell viability Artemisinine exerts its biological activity primarily through interaction with parasite-derived heme groups leading to the formation of reactive oxygen species that disrupt cell integrity and by inducing oxidative stress and apoptotic pathways in tumor cells In in vitro studies Artemisinine demonstrates cytotoxicity toward cancer cells and antimalarial activity with reported IC50 values ranging from 5 to 20 M depending on experimental conditions cell or parasite lines utilized Based on these pharmacological properties Artemisinine holds research potential in antimalarial and antitumor applications
Catalog No. 50-000-90369 Supplier Apexbio Technology LLC Supplier No. N1827200
Artemisinine (CAS 63968-64-9) is a sesquiterpene lactone designed to modulate parasite cell integrity and cancer cell viability Artemisinine exerts its biological activity primarily through interaction with parasite-derived heme groups leading to the formation of reactive oxygen species that disrupt cell integrity and by inducing oxidative stress and apoptotic pathways in tumor cells In in vitro studies Artemisinine demonstrates cytotoxicity toward cancer cells and antimalarial activity with reported IC50 values ranging from 5 to 20 M depending on experimental conditions cell or parasite lines utilized Based on these pharmacological properties Artemisinine holds research potential in antimalarial and antitumor applications