Learn More
TARGETMOL CHEMICALS INC ARTEMITIN 10MG
SDP
Catalog No. 502578913
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Artemitin (Erianthin) has anti-inflammatory activity. 2. Artemitin (Erianthin) protects endothelial function by acting as antioxidant and antiapoptotic agent and through the activation of ERK1/2 and Akt. 3. Intravenous injection of Artemitin (Erianthin)(.75 mg/kg) significantly reduces the hypertensive response to angiotensin I while increases the average length of bradykinin-induced hypotension. purity: 99%
Catalog No. 50-257-8913 Supplier TARGETMOL CHEMICALS INC Supplier No. TCS170410MG
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Artemitin (Erianthin) has anti-inflammatory activity. 2. Artemitin (Erianthin) protects endothelial function by acting as antioxidant and antiapoptotic agent and through the activation of ERK1/2 and Akt. 3. Intravenous injection of Artemitin (Erianthin)(.75 mg/kg) significantly reduces the hypertensive response to angiotensin I while increases the average length of bradykinin-induced hypotension. purity: 99%