Learn More
Cayman Chemical Arteannuin B, Purity: ≥98%, CAS: 50906-56-4, 10 mg
SDP
Catalog No. 502948252
Encompass_Preferred
A precursor of artemisinin with diverse biological activities; active against a variety of bacteria and fungi in disk assays; reduces viral RNA levels in Vero E6 cells infected with SARS-CoV-2 (EC50 = 10.28 µM); cytotoxic to Ehrlich ascites tumor cells (IC50 = 47 µM)
Catalog No. 50-294-8252 Supplier Cayman Chemical Supplier No. 3385210 MG
A precursor of artemisinin with diverse biological activities; active against a variety of bacteria and fungi in disk assays; reduces viral RNA levels in Vero E6 cells infected with SARS-CoV-2 (EC50 = 10.28 µM); cytotoxic to Ehrlich ascites tumor cells (IC50 = 47 µM)