Learn More
Medchemexpress LLC HY-10852 10mg Medchemexpress, Arterolane CAS:664338-39-0 Purity:>98%
SDP
Catalog No. 501968567
Encompass_Preferred
Medchemexpress, HY-10852 10mg Arterolane CAS:664338-39-0 Arterolane is an antimalarial agent, with IC50 of both 1.1 nM against P. falciparum Ro73 and W2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8567 Supplier Medchemexpress LLC Supplier No. HY1085210MG
Medchemexpress, HY-10852 10mg Arterolane CAS:664338-39-0 Arterolane is an antimalarial agent, with IC50 of both 1.1 nM against P. falciparum Ro73 and W2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.