Learn More
AdipoGen Artesunate (100 mg)
SDP
Supplier:  AdipoGen AGCN20469M100
Encompass_Preferred
Chemical. CAS: 88495-63-0. Formula: C19H28O8. MW: 384.4. Semi-synthetic derivative of artemisinin originally isolated from Artemisia annua. Semi-synthetic derivative of artemisinin. Anti-malaria. Anti-schistosomiasis. Inhibitor of human cytomegalovirus (CMV). Apoptosis inducer. Anti-cancer. Immunosuppressant. Inhibits production of tumor necrosis factor-alpha (TNF-alpha), interleukin-6 (IL-6), monocyte chemotactic protein-1 (MCP-1) and nitric oxide (NO).
Catalog No. 50-205-3649