Learn More
Abcam Artesunate, artemisinin derivative, 100MG
Catalog No. 502706166
Encompass_Preferred
MW 384.4 Da, Purity >98%. Semi-synthetic artemisinin (Asc -1308 ) derivative. Induces apoptosis of vascular endothelial cells. Shows antiangiogenenic and antiparasitaemic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6166 Supplier Abcam Supplier No. AB142829100MG
MW 384.4 Da, Purity >98%. Semi-synthetic artemisinin (Asc -1308 ) derivative. Induces apoptosis of vascular endothelial cells. Shows antiangiogenenic and antiparasitaemic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement