Learn More
Cayman Chemical Artesunate-d4 500ug
SDP
Catalog No. 502938973
Encompass_Preferred
An internal standard for the quantification of artesunate by GC- or LC-MS
Catalog No. 50-293-8973 Supplier Cayman Chemical Supplier No. 28769500 UG
An internal standard for the quantification of artesunate by GC- or LC-MS