Learn More
Pickering Laboratories Inc Artifical Saliva, Medical and Dental Research, Stabilized, each (200 mL/bottle)
SDP
Catalog No. NC1306283
Encompass_Preferred
This Artificial Saliva is formulated according to literature references for medical and dental research. This formulation has similar composition to commercially available products used to treat dry mouth and other conditions. This ready-to-use solution contains Sodium Carboxymethyl Cellulose to increase viscosity of the solution and make it behave similar to natural human saliva. This formulation contains a preservative and can be stored at room temperature and has pH of 6.8. This solution is only intended for product testing and research, and not for medical use
Catalog No. NC1306283 Supplier Pickering Laboratories Inc Supplier No. 17000305
This Artificial Saliva is formulated according to literature references for medical and dental research. This formulation has similar composition to commercially available products used to treat dry mouth and other conditions. This ready-to-use solution contains Sodium Carboxymethyl Cellulose to increase viscosity of the solution and make it behave similar to natural human saliva. This formulation contains a preservative and can be stored at room temperature and has pH of 6.8. This solution is only intended for product testing and research, and not for medical use