Learn More
HIMEDIA Artificial Sea Water Salts Broth, 500g
Catalog No. NC3595518
Encompass_Preferred
Artificial Sea Water Salts Broth is recommended for culturing marine bacteria. It is primarily used in marine biology and allows the easy preparation of media appropriate for marine organisms. Marine artificial media are used when critical studies cannot be conducted using a natural seawater base, so artificial seawater medium is used to minimize or exclude known contaminants for the purpose of studying trace elements.
Catalog No. NC3595518 Supplier HIMEDIA Supplier No. M1942500G
Artificial Sea Water Salts Broth is recommended for culturing marine bacteria. It is primarily used in marine biology and allows the easy preparation of media appropriate for marine organisms. Marine artificial media are used when critical studies cannot be conducted using a natural seawater base, so artificial seawater medium is used to minimize or exclude known contaminants for the purpose of studying trace elements.