Learn More
Med Vet International Arthroscopic Coronoid Process Osteotome, J1068
Catalog No. 5000585733
Encompass_Preferred
A comfortable fiber composite handle with metal end to take hammering. 1.1mm thick blade. This makes a great choice for procedures like block sulcoplasty. Blades can be changed intra-operatively for proper fit during the procedure. Supplied as a complete kit with 7 different blades, or purchase handle and blades separately.
Catalog No. 50-005-85733 Supplier Med Vet International Supplier No. JOR1068
A comfortable fiber composite handle with metal end to take hammering. 1.1mm thick blade. This makes a great choice for procedures like block sulcoplasty. Blades can be changed intra-operatively for proper fit during the procedure. Supplied as a complete kit with 7 different blades, or purchase handle and blades separately.