Learn More
Apexbio Technology LLC Articaine HCl 23964-57-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092276
Encompass_Preferred
Articaine hydrochloride (CAS 23964-57-0) is an amide-type local anesthetic widely utilized in dental research Structurally it incorporates an additional ester group which facilitates rapid hydrolysis by plasma and tissue esterases distinguishing its metabolic pathway from other local anesthetics Articaine exerts its action by inhibiting voltage-gated sodium channels thereby blocking nerve impulse conduction Due to its fast onset and intermediate duration of action Articaine HCl is employed in studies investigating neural conduction pain modulation and comparative pharmacokinetics of local anesthetic agents in preclinical models
Catalog No. 50-000-92276 Supplier Apexbio Technology LLC Supplier No. B18875.1
Articaine hydrochloride (CAS 23964-57-0) is an amide-type local anesthetic widely utilized in dental research Structurally it incorporates an additional ester group which facilitates rapid hydrolysis by plasma and tissue esterases distinguishing its metabolic pathway from other local anesthetics Articaine exerts its action by inhibiting voltage-gated sodium channels thereby blocking nerve impulse conduction Due to its fast onset and intermediate duration of action Articaine HCl is employed in studies investigating neural conduction pain modulation and comparative pharmacokinetics of local anesthetic agents in preclinical models