Learn More
Medchemexpress LLC Articaine (hydrochloride) | 23964-57-0 | 99.38% | 25 MG
SDP
Catalog No. 5000450654
Encompass_Preferred
Articaine hydrochloride is a selective inhibitor of voltage-gated sodium channels (e.g., rNav1.4, hNav1.7, and rNav1.8). It exhibits local anesthetic activity by inhibiting Na+ influx to block nerve impulse conduction. Additionally, it can inhibit NF-κB activation and NLRP3 inflammasome pathways, demonstrating anti-inflammatory properties. This compound is suitable for studying dental local anesthesia and inflammatory-related diseases such as acute kidney injury.
  • Selective inhibitor of voltage-gated sodium channels
  • Exhibits local anesthetic activity by inhibiting Na+ influx
  • Can inhibit NF-κB activation and NLRP3 inflammasome pathways
  • Displays anti-inflammatory function
  • Used in research for dental local anesthesia and inflammatory-related diseases
Catalog No. 50-004-50654 Supplier Medchemexpress LLC Supplier No. 25MGHYB0516
Articaine hydrochloride is a selective inhibitor of voltage-gated sodium channels (e.g., rNav1.4, hNav1.7, and rNav1.8). It exhibits local anesthetic activity by inhibiting Na+ influx to block nerve impulse conduction. Additionally, it can inhibit NF-κB activation and NLRP3 inflammasome pathways, demonstrating anti-inflammatory properties. This compound is suitable for studying dental local anesthesia and inflammatory-related diseases such as acute kidney injury.
  • Selective inhibitor of voltage-gated sodium channels
  • Exhibits local anesthetic activity by inhibiting Na+ influx
  • Can inhibit NF-κB activation and NLRP3 inflammasome pathways
  • Displays anti-inflammatory function
  • Used in research for dental local anesthesia and inflammatory-related diseases