Learn More
Medchemexpress LLC Articaine hydrochloride | 23964-57-0 | 99.4% | 1 ML
SDP
Catalog No. 5000407599
Encompass_Preferred
Articaine hydrochloride is a selective inhibitor of voltage-gated sodium channels, demonstrating local anesthetic activity by blocking nerve impulse conduction. It also exhibits anti-inflammatory functions by inhibiting NF-κB activation and NLRP3 inflammasome pathways. It is used in the study of dental local anesthesia and inflammatory-related diseases.
  • Inhibits voltage-gated sodium channels
  • Exhibits local anesthetic activity
  • Inhibits NF-κB activation and NLRP3 inflammasome pathways
  • Reduces inflammatory factors like TNF-α, IL-6, and IL-1β
  • Ameliorates LPS-induced acute kidney injury in animal models
  • Can enhance anesthetic effect in zebrafish model
Catalog No. 50-004-07599 Supplier Medchemexpress LLC Supplier No. HYB051610MM/1ML
Articaine hydrochloride is a selective inhibitor of voltage-gated sodium channels, demonstrating local anesthetic activity by blocking nerve impulse conduction. It also exhibits anti-inflammatory functions by inhibiting NF-κB activation and NLRP3 inflammasome pathways. It is used in the study of dental local anesthesia and inflammatory-related diseases.
  • Inhibits voltage-gated sodium channels
  • Exhibits local anesthetic activity
  • Inhibits NF-κB activation and NLRP3 inflammasome pathways
  • Reduces inflammatory factors like TNF-α, IL-6, and IL-1β
  • Ameliorates LPS-induced acute kidney injury in animal models
  • Can enhance anesthetic effect in zebrafish model