Learn More
Cayman Chemical Articaine (hydrochloride), Purity: ≥98%, CAS: 23964-57-0, 100 mg
SDP
Catalog No. 502965138
Encompass_Preferred
An amide local anesthetic most commonly used in dentistry; it possesses a unique ester side chain that is metabolized rapidly by circulating esterases
Catalog No. 50-296-5138 Supplier Cayman Chemical Supplier No. 17883100 MG
An amide local anesthetic most commonly used in dentistry; it possesses a unique ester side chain that is metabolized rapidly by circulating esterases