Learn More
Med Vet International Miltex Articulate Paper, 12 Packs, 528-50750
Catalog No. 502831852
Encompass_Preferred
Articulating paper is used to mark and identify contact points between the maxillary and mandibular teeth during all forms of natural tooth occlusal adjustments and dental prosthesis insertions. 12 Pack.
Catalog No. 50-283-1852 Supplier Med Vet International Supplier No. MIL52850750
Articulating paper is used to mark and identify contact points between the maxillary and mandibular teeth during all forms of natural tooth occlusal adjustments and dental prosthesis insertions. 12 Pack.