Learn More
World Precision Instrument Articulated Arm and Table Clamp
SDP
Catalog No. 502521350
Encompass
The small horizontal bar is always parallel to the table surface. The small horizontal arm is permanently linked with the adjustable forearm, and the two are raised and lowered together. Within the adjustable forearm is an internal tension spring to prevent drift. The unit has three pivot points of rotation, and all have a manual clutch allowing you to lock in the position:
Catalog No. 50-252-1350 Supplier World Precision Instrument Supplier No. 502007
The small horizontal bar is always parallel to the table surface. The small horizontal arm is permanently linked with the adjustable forearm, and the two are raised and lowered together. Within the adjustable forearm is an internal tension spring to prevent drift. The unit has three pivot points of rotation, and all have a manual clutch allowing you to lock in the position: