Learn More
Pickering Laboratories Inc Artificial Eccrine Perspiration , Stabilized, each (5 mL/bottle)
SDP
Catalog No. NC0328484
Encompass_Preferred
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. Pickering Laboratories’ human mimic artificial perspiration solutions are formulated based on published data for human sweat and provide the closest match to the “real thing” available on the market. These solutions are perfect for testing a variety of consumer products and medical devices as well as for forensics applications. Artificial Eccrine Perspiration is a ready to-use human eccrine sweat solution. It consists of nineteen amino acids, the seven most abundant minerals, and the four most abundant metabolites at a pH of 4.5.
Catalog No. NC0328484 Supplier Pickering Laboratories Inc Supplier No. 17000024
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. Pickering Laboratories’ human mimic artificial perspiration solutions are formulated based on published data for human sweat and provide the closest match to the “real thing” available on the market. These solutions are perfect for testing a variety of consumer products and medical devices as well as for forensics applications. Artificial Eccrine Perspiration is a ready to-use human eccrine sweat solution. It consists of nineteen amino acids, the seven most abundant minerals, and the four most abundant metabolites at a pH of 4.5.