Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Artificial Blood
SDPSCI_ED

Catalog No. S00283 Shop All 3B Scientific Products
Change view
Click to view available options
Size:
1 L
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S00283 1 L
S00284 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S00283 Supplier 3B Scientific™ Supplier No. 1005611
Add to Cart
Add to Cart
This item is not returnable. View return policy

Although not the same viscosity as real blood, Life/form™ simulation blood provides a sense of reality to any activity.

  • Although not the same viscosity as real blood, Life/form™ simulation blood provides a sense of reality to any activity.
  • For use with Life/form™ Products:
    • Advanced Venipuncture and Injection Arm (W44216, W44217)
    • Venipuncture and Injection Demonstration Arm
    • Deluxe IV Training Arm
    • Venipuncture Training Arm
    • Pediatric Arm (W44715)
    • Pediatric Head (W44101)
    • Central Venous Cannulation Simulator (W44017)
    • Heart Catherization Simulator for Total Parenteral Nutrition and Central Venous Catheterization (W44015)
    • First Aid Arm (W44733)

Specifications

Product Type Artificial Blood
Color Red
For Use With (Equipment) Life/Form™ Advanced Venipuncture and Injection Arm, Venipuncture and Injection Demonstration Arm, Deluxe IV Training Arm, Venipuncture Training Arm, Pediatric Arm, Pediatric Head, Central Venous Cannulation Simulator, Heart Catherization Simulator for Total Parenteral Nutrition and Central Venous Catheterization, First Aid Arm
Size 1 L
Weight (English) 0.18 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.