Learn More
Pickering Laboratories Inc Artificial Perspiration, ISO 3160, Stabilized, each (19.8L/carboy)
SDP
Catalog No. 501974220
Encompass_Preferred
This formulation is used to determine corrosion (tarnishing, oxidation and surface penetration) resistance for gold alloy coverings on watch cases and accessories, including bracelets when they are permanently attached to the case. The solution is at pH 4.7 per ISO 3160 specifications
Catalog No. 50-197-4220 Supplier Pickering Laboratories Inc Supplier No. 17000557
This formulation is used to determine corrosion (tarnishing, oxidation and surface penetration) resistance for gold alloy coverings on watch cases and accessories, including bracelets when they are permanently attached to the case. The solution is at pH 4.7 per ISO 3160 specifications