Learn More
Pickering Laboratories Inc Artificial Perspiration, BS EN 1811:2011, Stabilized, each (19.8 L/carboy)
SDP
Catalog No. 501974202
Encompass_Preferred
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. This pH 6.5 formulation is used to stimulate the release of Nickel from all post assemblies which are inserted into pierced ears and other pierced parts of the human body, and for articles intended to come into direct and prolonged contact with the skin.
Catalog No. 50-197-4202 Supplier Pickering Laboratories Inc Supplier No. 17000566
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. This pH 6.5 formulation is used to stimulate the release of Nickel from all post assemblies which are inserted into pierced ears and other pierced parts of the human body, and for articles intended to come into direct and prolonged contact with the skin.