Learn More
Pickering Laboratories Inc Artificial Perspiration, ISO 3160, Custom pH, Not Stabilized, each (200 mL /bottle)
SDP
Catalog No. 501974217
Encompass_Preferred
This formulation is used to determine corrosion (tarnishing, oxidation and surface penetration) resistance for gold alloy coverings on watch cases and accessories, including bracelets when they are permanently attached to the case. Provided at a pH level of your choosing
Catalog No. 50-197-4217 Supplier Pickering Laboratories Inc Supplier No. 17000520
Add to Cart
Add to Cart
This formulation is used to determine corrosion (tarnishing, oxidation and surface penetration) resistance for gold alloy coverings on watch cases and accessories, including bracelets when they are permanently attached to the case. Provided at a pH level of your choosing