Learn More
Pickering Laboratories Inc Artificial Perspiration, DIN-EN/IEC60068-2-70, Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501974204
Encompass_Preferred
This formulation is used to determine the resistance of markings and letterings on flat or curved surfaces against abrasion as it may occur by manually operating actuators and keyboards. This formulation can also be used to determine resistance against fluid contamination as it may occur under normal use. Designed to meet DIN EN 60068-2-70 & IEC 60068-2-70 regulations
Catalog No. 50-197-4204 Supplier Pickering Laboratories Inc Supplier No. 17000543
This formulation is used to determine the resistance of markings and letterings on flat or curved surfaces against abrasion as it may occur by manually operating actuators and keyboards. This formulation can also be used to determine resistance against fluid contamination as it may occur under normal use. Designed to meet DIN EN 60068-2-70 & IEC 60068-2-70 regulations