Learn More
Pickering Laboratories Inc Artificial Perspiration, ISO 3160, Custom pH, Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501974219
Encompass_Preferred
This formulation is used to determine corrosion (tarnishing, oxidation and surface penetration) resistance for gold alloy coverings on watch cases and accessories, including bracelets when they are permanently attached to the case. The solution is at pH of your choosing
Catalog No. 50-197-4219 Supplier Pickering Laboratories Inc Supplier No. 17000526
This formulation is used to determine corrosion (tarnishing, oxidation and surface penetration) resistance for gold alloy coverings on watch cases and accessories, including bracelets when they are permanently attached to the case. The solution is at pH of your choosing