Learn More
Pickering Laboratories Inc Artificial Perspiration, BS EN 1811:2011, Custom pH, Stabilized, each (200 mL /bottle)
SDP
Supplier:  Pickering Laboratories Inc 17000515
Encompass_Preferred
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. This formulation is used to stimulate the release of Nickel from all post assemblies which are inserted into pierced ears and other pierced parts of the human body, and for articles intended to come into direct and prolonged contact with the skin. Provided at a pH of your choosing
Catalog No. 50-197-4199