Learn More
Pickering Laboratories Inc Artificial Perspiration, ASTM D2322-00, Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501974197
Encompass_Preferred
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. ASTM method D2322-00 determines the resistance to grain cracking, area loss, and flexibility of shoe upper leather to artificial perspiration
Catalog No. 50-197-4197 Supplier Pickering Laboratories Inc Supplier No. 17000550
Add to Cart
Add to Cart
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. ASTM method D2322-00 determines the resistance to grain cracking, area loss, and flexibility of shoe upper leather to artificial perspiration