Learn More
Pickering Laboratories Inc Artificial Perspiration, ISO 11641, Custom pH, Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501974211
Encompass_Preferred
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. This formulation, per ISO 11641, determines colorfastness to perspiration of leather at all stages of processing, particularly to gloving, clothing and lining leathers, and uppers of unlined shoes. This alkaline perspiration has a greater effect on the color of leather and is used to simulate the most demanding conditions. Provided at a pH level of your choosing
Catalog No. 50-197-4211 Supplier Pickering Laboratories Inc Supplier No. 17000524
Add to Cart
Add to Cart
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. This formulation, per ISO 11641, determines colorfastness to perspiration of leather at all stages of processing, particularly to gloving, clothing and lining leathers, and uppers of unlined shoes. This alkaline perspiration has a greater effect on the color of leather and is used to simulate the most demanding conditions. Provided at a pH level of your choosing