Learn More
Pickering Laboratories Inc Artificial Perspiration, ISO 3160, Custom pH, Stabilized, each (950 mL/bottle)
SDP
Catalog No. 501974218
Encompass_Preferred
This formulation is used to determine corrosion (tarnishing, oxidation and surface penetration) resistance for gold alloy coverings on watch cases and accessories, including bracelets when they are permanently attached to the case. Provided at a pH level of your choosing
Catalog No. 50-197-4218 Supplier Pickering Laboratories Inc Supplier No. 17000545
This formulation is used to determine corrosion (tarnishing, oxidation and surface penetration) resistance for gold alloy coverings on watch cases and accessories, including bracelets when they are permanently attached to the case. Provided at a pH level of your choosing