Learn More
Pickering Laboratories Inc Artificial Perspiration, ISO 3160 with added Pyruvic acid, Buffered, Stabilized, each (950 mL/bottle).
SDP
Catalog No. NC1873823
Encompass_Preferred
This formulation is used to determine corrosion (tarnishing, oxidation and surface penetration) resistance for gold alloy coverings on watch cases and accessories, including bracelets when they are permanently attached to the case. This formulation has an addition of Pyruvic Acid and Phosphate Buffer for added pH stability
Catalog No. NC1873823 Supplier Pickering Laboratories Inc Supplier No. 17000537
This formulation is used to determine corrosion (tarnishing, oxidation and surface penetration) resistance for gold alloy coverings on watch cases and accessories, including bracelets when they are permanently attached to the case. This formulation has an addition of Pyruvic Acid and Phosphate Buffer for added pH stability