Learn More
Pickering Laboratories Inc Artificial Saliva, Pharmaceutical Research, Not Stabilized, each (19.8 L/carboy)
SDP
Catalog No. 501974222
Encompass_Preferred
Artificial Saliva is formulated according to literature for pharmaceutical research such as studies of drug dissolution and drug delivery through oral mucosa. This is a ready to use formulation that should be stored refrigerated. The pH of the solution is 6.8
Catalog No. 50-197-4222 Supplier Pickering Laboratories Inc Supplier No. 17000355
Artificial Saliva is formulated according to literature for pharmaceutical research such as studies of drug dissolution and drug delivery through oral mucosa. This is a ready to use formulation that should be stored refrigerated. The pH of the solution is 6.8