Learn More
Pickering Laboratories Inc Artificial Saliva, Fusayama/Mayer, Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501974226
Encompass_Preferred
This ready-to-use solution closely resembles the mineral composition of natural saliva and can be used for testing a wide variety of products, including dental metal alloys. This formulation is at pH 4.9 and should be stored refrigerated.
Catalog No. 50-197-4226 Supplier Pickering Laboratories Inc Supplier No. 17000309
This ready-to-use solution closely resembles the mineral composition of natural saliva and can be used for testing a wide variety of products, including dental metal alloys. This formulation is at pH 4.9 and should be stored refrigerated.