Learn More
Pickering Laboratories Inc Artificial Saliva, Pharmaceutical Research, Custom pH, Not Stabilized, each (200mL/bottle)
SDP
Catalog No. 501974229
Encompass_Preferred
Artificial Saliva is formulated according to literature for pharmaceutical research such as studies of drug dissolution and drug delivery through oral mucosa. This is a ready to use formulation that should be stored refrigerated. Provided at a pH level of your choosing
Catalog No. 50-197-4229 Supplier Pickering Laboratories Inc Supplier No. 17000308
Artificial Saliva is formulated according to literature for pharmaceutical research such as studies of drug dissolution and drug delivery through oral mucosa. This is a ready to use formulation that should be stored refrigerated. Provided at a pH level of your choosing