Learn More
Pickering Laboratories Inc Artificial Saliva, Fusayama/Mayer, Custom pH, Not Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501974224
Encompass_Preferred
This ready-to-use solution closely resembles themineral composition of natural saliva and can be usedfor testing a wide variety of products, including dentalmetal alloys. This formulation is at pH 4.9 and shouldbe stored refrigerated.
Catalog No. 50-197-4224 Supplier Pickering Laboratories Inc Supplier No. 17000307
This ready-to-use solution closely resembles themineral composition of natural saliva and can be usedfor testing a wide variety of products, including dentalmetal alloys. This formulation is at pH 4.9 and shouldbe stored refrigerated.