Learn More
Pickering Laboratories Inc Artificial Saliva, ISO 10271-2011, Not Stabilzied, each (200 mL/bottle)
SDP
Catalog No. 501974227
Encompass_Preferred
This Artificial Saliva is formulated according to ISO 10271-2011 standard procedure. ISO 10271-2011 specifies the method for corrosion testing for metallic material in dental devices. The solution should be stored in the refrigerator. The pH of the solution is 2.3.
Catalog No. 50-197-4227 Supplier Pickering Laboratories Inc Supplier No. 17000313
This Artificial Saliva is formulated according to ISO 10271-2011 standard procedure. ISO 10271-2011 specifies the method for corrosion testing for metallic material in dental devices. The solution should be stored in the refrigerator. The pH of the solution is 2.3.