Learn More
Pickering Laboratories Inc Artificial Saliva, Fusayama/Mayer, Custom pH, Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501974225
Encompass_Preferred
This ready-to-use solution closely resembles themineral composition of natural saliva and can be used for testing a wide variety of products, including dental metal alloys. This formulation is at pH 4.9 and should be stored refrigerated.
Catalog No. 50-197-4225 Supplier Pickering Laboratories Inc Supplier No. 17000306
This ready-to-use solution closely resembles themineral composition of natural saliva and can be used for testing a wide variety of products, including dental metal alloys. This formulation is at pH 4.9 and should be stored refrigerated.