Learn More
Pickering Laboratories Inc Artificial Saliva, ASTM E2720-16 and ASTM E2721-16 with mucin, Stabilized, each (200 mL/bottle).
SDP
Catalog No. NC1873811
Encompass_Preferred
This artificial saliva formulation has a mineral composition and pH close to human saliva. Mucin is added to increase the viscosity and lubricating ability of the artificial saliva. This solution can be used for dental, drug delivery or pharmaceutical studies. The stabilized solution is pre-mixed and has a preservative added to prevent bacterial growth; it is stored at room temperature. The pH of the formulation is 6.8. Amylase is the main enzyme in human saliva and is needed for starch digestion. As some applications require studying the effect of enzymes in saliva, Pickering Laboratories offers an easy way to add biological activity to your chosen artificial saliva product. Once added to the 200 mL bottle of artificial saliva, our Amylase product will provide 80 U/mL of enzyme activity
Catalog No. NC1873811 Supplier Pickering Laboratories Inc Supplier No. 17000316
This artificial saliva formulation has a mineral composition and pH close to human saliva. Mucin is added to increase the viscosity and lubricating ability of the artificial saliva. This solution can be used for dental, drug delivery or pharmaceutical studies. The stabilized solution is pre-mixed and has a preservative added to prevent bacterial growth; it is stored at room temperature. The pH of the formulation is 6.8. Amylase is the main enzyme in human saliva and is needed for starch digestion. As some applications require studying the effect of enzymes in saliva, Pickering Laboratories offers an easy way to add biological activity to your chosen artificial saliva product. Once added to the 200 mL bottle of artificial saliva, our Amylase product will provide 80 U/mL of enzyme activity