Learn More
Pickering Laboratories Inc Artificial Saliva, DIN 53160, Not Stabilized, Pre-mixed, 1 month shelf life, each (200 mL/bottle)
SDP
Catalog No. NC1873820
Encompass_Preferred
Artificial saliva is prepared according to specifications of DIN 53160-1:200, SN/T 1058.1-2013 and GB/T 18886-2019 official methods. The pH of the solution after mixing is 6.8. Pre-mixed with a 1 month shelf life. Made to order
Catalog No. NC1873820 Supplier Pickering Laboratories Inc Supplier No. 17000323
Artificial saliva is prepared according to specifications of DIN 53160-1:200, SN/T 1058.1-2013 and GB/T 18886-2019 official methods. The pH of the solution after mixing is 6.8. Pre-mixed with a 1 month shelf life. Made to order